Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ p14 ARF/p16β Ab-2, Mouse Monoclonal Antibody

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ p14ARF/p16β Ab-2, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS850P1

 View more versions of this product

Catalog No. MS850P1

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ p14ARF/p16β Ab-2, Mouse Monoclonal Antibody.

The INK4a-ARF locus encodes two unrelated proteins both of which function in tumor suppression. p14ARF arrests the cell cycle in a p53-dependent manner. p14ARF binds to mdm2 and promotes the rapid degradation of mdm2 protein required for mdm2 modification and concurrent p53 stabilization and accumulation. This interaction is mediated by the exon 1Β-encoded N-terminal domain of p14ARF and a C-terminal region of mdm2. Deletion of the ARF-INK4a locus simultaneously impairs both the INK4a-cyclin D/cdk4-RB and the ARF-mdm2-p53 pathways.

Host Species: Mouse

Clone: 14P02; same as DCS-240

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: N-terminal

Immunogen: Recombinant p14ARF protein

Molecular Weight: 14kDa

Positive Control: HT-29 or HeLa cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Not optimum)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

p14ARF/p16beta Ab-2
Monoclonal
200μg/mL
Concentrated
Recombinant p14ARF protein
500μL
Cell Cycle
Primary
Human
Immunofluorescence, Western Blot
14P02; same as DCS-240
Unconjugated
Murine
IgG2a kappa
RUO
Primary
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit