Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p15INK4b/CDKN2B Mouse anti-Human, Clone: DCS114.1, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266636
Description
p15INK4b/CDKN2B Monoclonal antibody specifically detects p15INK4b/CDKN2B in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
p15INK4b/CDKN2B | |
DCS114.1 | |
Western Blot, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500 | |
Purified | |
CDKN2B | |
A his-tagged recombinant full length p15 protein recognizing a 15kDa protein | |
Protein A or G purified | |
Cell Cycle and Replication | |
Primary | |
1030 |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
PBS with 0.08% Sodium Azide | |
CDK4I, cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4), cyclin-dependent kinases 4 and 6 binding protein, INK4BCDK4B inhibitor, MTS-2, MTS2CDK inhibitory protein, Multiple tumor suppressor 2, p14_CDK inhibitor, p14_INK4B, p14-INK4b, p15 CDK inhibitor, p15_INK4B, P15cyclin-dependent kinase 4 inhibitor B, p15INK4b, p15-INK4b, TP15 | |
Mouse | |
IgG1 | |
100μg | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title