Learn More
p18INK4c Ab-3 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results with the Epredia™ p18INK4c Ab-3, Mouse Monoclonal Antibody.
Supplier: Epredia MS858P1ABX
Description
p18INK4c is a specific inhibitor of cdk4/cdk6 and is expressed during G1 -to S transition in the eukaryotic cell division cycle. It is maximally induced as cells enter S-phase.Host Species: Mouse
Clone: 18P118; same as DCS-118
Isotype: IgG1
Species Reactivity: Human, Mouse. Others not known.
Epitope: Not determined
Immunogen: Purified recombinant p18INK4c protein
Molecular Weight: 18kDa
Positive Control: HeLa cells; Tonsil, skeletal muscle, heart, pancreas
Cellular Localization: Cytoplasmic and Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation
Note:
USA: RUO; Int'l: RUO
Specifications
p18superscriptINK4csuperscript Ab-3 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Cell Cycle | |
Human, Mouse | |
IgG1 |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
18P118; same as DCS-118 | |
Unconjugated | |
Purified recombinant p18superscriptINK4csuperscript protein | |
RUO | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.