Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p18INK4c Ab-3 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results with the Epredia™ p18INK4c Ab-3, Mouse Monoclonal Antibody.

Supplier:  Epredia MS858P

 View more versions of this product

Catalog No. MS858P

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

p18INK4c is a specific inhibitor of cdk4/cdk6 and is expressed during G1 -to S transition in the eukaryotic cell division cycle. It is maximally induced as cells enter S-phase.

Host Species: Mouse

Clone: 18P118; same as DCS-118

Isotype: IgG1

Species Reactivity: Human, Mouse. Others not known.

Epitope: Not determined

Immunogen: Purified recombinant p18INK4c protein

Molecular Weight: 18kDa

Positive Control: HeLa cells; Tonsil, skeletal muscle, heart, pancreas

Cellular Localization: Cytoplasmic and Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

p18superscriptINK4csuperscript Ab-3
Monoclonal
200μg/mL
Mouse
1 mL
Cell Cycle
Human, Mouse
IgG1
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
18P118; same as DCS-118
Unconjugated
Purified recombinant p18superscriptINK4csuperscript protein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit