Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ p18 INK4c Ab-3, Mouse Monoclonal Antibody

Ensure accurate, reproducible results with the Thermo Scientific™ p18INK4c Ab-3, Mouse Monoclonal Antibody.

$174.00 - $807.00

Specifications

Antigen p18superscriptINK4csuperscript Ab-3
Clone 18P118; same as DCS-118
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 7
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS858P
View Documents
Thermo Scientific™
MS858P
Purified Ab with BSA and Azide 1mL
Each for $741.50
Add to cart
 
MS858P0
View Documents
Thermo Scientific™
MS858P0
Purified Ab with BSA and Azide 100μL
Each for $174.00
Add to cart
 
MS858P1
View Documents
Thermo Scientific™
MS858P1
Purified Ab with BSA and Azide 500μL
Each for $487.00
Add to cart
 
MS858P1ABX
View Documents
Thermo Scientific™
MS858P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $541.50
Add to cart
 
MS858PABX
View Documents
Thermo Scientific™
MS858PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $807.00
Add to cart
 
MS858R7
View Documents
Thermo Scientific™
MS858R7
Ready-to-use for IHC (UVLP) 7mL
Each for $524.50
Add to cart
 
MS858R1
View Documents
Thermo Scientific™
MS858R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Ensure accurate, reproducible results with the Thermo Scientific™ p18INK4c Ab-3, Mouse Monoclonal Antibody.

p18INK4c is a specific inhibitor of cdk4/cdk6 and is expressed during G1 -to S transition in the eukaryotic cell division cycle. It is maximally induced as cells enter S-phase.

Host Species: Mouse

Clone: 18P118; same as DCS-118

Isotype: IgG1

Species Reactivity: Human, Mouse. Others not known.

Epitope: Not determined

Immunogen: Purified recombinant p18INK4c protein

Molecular Weight: 18kDa

Positive Control: HeLa cells; Tonsil, skeletal muscle, heart, pancreas

Cellular Localization: Cytoplasmic and Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

p18superscriptINK4csuperscript Ab-3
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RUO
Primary
IgG1
18P118; same as DCS-118
Monoclonal
Murine
Cell Cycle
Purified recombinant p18superscriptINK4csuperscript protein
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit