Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p18INK4c/CDKN2C Mouse anti-Human, Mouse, Rat, Biotin, Clone: OTI2E1, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273185B
Description
p18INK4c/CDKN2C Monoclonal specifically detects p18INK4c/CDKN2C in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
p18INK4c/CDKN2C | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
CDKN2C | |
Full length recombinant protein of human p18INK4c/CDKN2C (NP_001253) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot | |
OTI2E1 | |
Western Blot | |
CDK6 inhibitor p18, CDKN6, cyclin-dependent inhibitor, cyclin-dependent kinase 4 inhibitor C, Cyclin-dependent kinase 6 inhibitor, cyclin-dependent kinase inhibitor 2C (p18, inhibits CDK4), INK4Ccyclin-dependent kinase 6 inhibitor p18, p18, p18-INK4C, p18-INK6 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
1031.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title