Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p19 INK4d Mouse anti-Human, Clone: DCS100, Novus Biologicals™
Mouse Monoclonal Antibody
$216.50 - $458.00
Specifications
Antigen | p19 INK4d |
---|---|
Clone | DCS100 |
Dilution | Western Blot, Immunohistochemistry, Immunoprecipitation |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
Classification | Monoclonal |
Description
p19 INK4d Monoclonal antibody specifically detects p19 INK4d in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
p19 INK4d | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CDK inhibitor p19INK4d, cell cycle inhibitor, Nur77 associating protein, cyclin-dependent kinase 4 inhibitor D, cyclin-dependent kinase 4 inhibitor D p19, cyclin-dependent kinase inhibitor 2D (p19, inhibits CDK4), inhibitor of cyclin-dependent kinase 4d, INK4D, p19, p19-INK4D | |
A his-tagged recombinant full length p19 protein recognizing a 19kD protein (Uniprot: P55273) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
DCS100 | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Cell Cycle and Replication, Ovarian Carcinoma Cell Markers | |
PBS | |
1032 | |
IgG1 | |
Protein A or G purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title