Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ p19 INK4d Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA527704

Catalog No. PIPA527704


Only null left
Add to Cart

Description

Description

Recommended positive controls: Rat bone marrow. Predicted reactivity: Mouse (88%), Rat (88%), Rhesus Monkey (98%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

p19 is a member of the INK4 family of cyclin-dependent kinase inhibitors. This protein has been shown to form a stable complex with CDK4 or CDK6, and prevent the activation of the CDK kinases, thus function as a cell growth regulator that controls cell cycle G1 progression. The abundance of the transcript of this gene was found to oscillate in a cell-cycle dependent manner with the lowest expression at mid G1 and a maximal expression during S phase. The negative regulation of the cell cycle involved in this protein was shown to participate in repressing neuronal proliferation, as well as spermatogenesis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

p19 INK4d
Polyclonal
Unconjugated
CDKN2D
CDK inhibitor p19INK4d; CDK4I; CDKN2D; cell cycle inhibitor, Nur77 associating protein; cyclin dependent kinase inhibitor 2D; cyclin-dependent kinase 4 inhibitor D; cyclin-dependent kinase 4 inhibitor D p19; cyclin-dependent kinase inhibitor 2D (p19, inhibits CDK4); inhibitor of cyclin-dependent kinase 4d; INK4d; MTS1; p16-INK4; p16-INK4a; p19; p19INK4d; p19-INK4D; similar to cyclin-dependent kinase inhibitor 2D
Rabbit
Protein A
RUO
1032, 494444
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
2 mg/mL
PBS with 40% glycerol and 0.01% thimerosal; pH 7
P55273
CDKN2D
Recombinant fragment corresponding to a region within amino acids 1 and 165 of Human p19 INK4d.
100 μL
Primary
Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.