Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p190RhoGAP Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP180051

 View more versions of this product

Catalog No. NBP180051


Add to cart

Description

Description

p190RhoGAP Polyclonal antibody specifically detects p190RhoGAP in Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish samples. It is validated for Western Blot.
Specifications

Specifications

p190RhoGAP
Unconjugated
PBS & 2% Sucrose. with 0.09% Sodium Azide
glucocorticoid receptor DNA binding factor 1, glucocorticoid receptor DNA-binding factor 1, glucocorticoid receptor repression factor 1, GRF1, GRF-1, GRLF1, KIAA1722, MGC10745, P190A, P190-A, p190ARhoGAP, p190RhoGAP, rho GAP p190A, Rho GTPase activating protein 35, rho GTPase-activating protein 35
Rabbit
IgG
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Western Blot
Western Blot 1:1000
Affinity Purified
ARHGAP35
Synthetic peptide directed towards the N terminal of human GRLF1. Peptide sequence RPSADEFHLDHTSVLSTSDFGGRVVNNDHFLYWGEVSRSLEDCVECKMHI.
171 kDa
100 ul
Cell Cycle and Replication
Primary
2909
Bovine, Canine, Equine, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit