Learn More
Alfa Aesar™ p21-Activated Kinase Inhibitor III, IPA-3
Acts as an allosteric inhibitor of Pak1
Manufacturer:  Alfa Aesar™ J65848MA
 View more versions of this product
Catalog No. AAJ65848MA
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
10mg
−30° to −10°C
Acts as an allosteric inhibitor of Pak1
C20H14O2S2
PAK Inhibitor III; 1,1′-Disulfanediylbis(naphthalen-2-ol)
Soluble in DMSO or ethanol; Insoluble in water
Yellow
Solid
42521-82-4
350.50g/mol
Light Sensitive
Signal Transduction Reagent