Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p21/CIP1/CDKN1A Mouse anti-Human, Mouse, FITC, Clone: 1032, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243697F

Catalog No. NBP243697F


Add to cart

Description

Description

p21/CIP1/CDKN1A Monoclonal antibody specifically detects p21/CIP1/CDKN1A in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications

Specifications

p21/CIP1/CDKN1A
1032
Immunocytochemistry/Immunofluorescence, Knockout Validated
Purified
CDKN1A
Recombinant protein encompassing a sequence within the center region of human p21 Cip1. The exact sequence is proprietary.
Protein G purified
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair, Phospho Specific
Primary
1026
Immunocytochemistry, Immunofluorescence, Immunoassay
FITC
PBS with 0.05% Sodium Azide
CAP20cyclin-dependent kinase inhibitor 1, CDK-interacting protein 1, CDKN1melanoma differentiation associated protein 6, CIP1WAF1CDK-interaction protein 1, cyclin-dependent kinase inhibitor 1A (p21, Cip1), MDA6, MDA-6, Melanoma differentiation-associated protein 6, p21, p21CIP1, p21Cip1/Waf1, PIC1, SDI1DNA synthesis inhibitor, wild-type p53-activated fragment 1
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit