Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p21/CIP1/CDKN1A Antibody (CIP1/2275R), Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP280488
Description
p21/CIP1/CDKN1A Monoclonal specifically detects p21/CIP1/CDKN1A in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
p21/CIP1/CDKN1A | |
Monoclonal | |
0.2 mg/mL | |
Western Blot 1 - 2 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin | |
P38936 | |
CDKN1A | |
Recombinant full-length human p21/CIP1/CDKN1A protein (Uniprot: P38936) | |
20 μg | |
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair, Phospho Specific | |
1026 | |
Human, Mouse (Negative), Rat (Negative) | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
CIP1/2275R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
CAP20cyclin-dependent kinase inhibitor 1, CDK-interacting protein 1, CDKN1melanoma differentiation associated protein 6, CIP1WAF1CDK-interaction protein 1, cyclin-dependent kinase inhibitor 1A (p21, Cip1), MDA6, MDA-6, Melanoma differentiation-associated protein 6, p21, p21CIP1, p21Cip1/Waf1, PIC1, SDI1DNA synthesis inhibitor, wild-type p53-activated fragment 1 | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
This monoclonal antibody recognizes a 21kDa protein, identified as the p21WAF1 tumor suppressor protein. This monoclonal antibody is highly specific to p21 and shows no cross-reaction with other closely related mitotic inhibitors. p21WAF1 is a specific inhibitor of cdks and a tumor suppressor involved in the pathogenesis of a variety of malignancies. The expression of this gene acts as an inhibitor of the cell cycle during G1 phase and is tightly controlled by the tumor suppressor protein p53. Its expression is induced by the wild type, but not mutant, p53 suppressor protein. Normal cells generally display a rather intense nuclear p21 expression. Loss of p21 expression has been reported in many carcinomas (gastric carcinoma, non-small cell lung carcinoma, thyroid carcinoma). | |
Store at 4°C. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction