Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p21 Rabbit anti-Human,Mouse, Unconjugated, Clone: 5G7, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen p21
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB114125MI
View Documents
MilliporeSigma
ZRB114125UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB11414XMI
View Documents
MilliporeSigma
ZRB11414X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

p21 Monoclonal antibody specifically detects p21 in Human,Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

p21
Unconjugated
Rabbit
Cyclin-dependent kinase inhibitor 1;CDK-interacting protein 1;Melanoma differentiation-associated protein 6;MDA-6;p21
His-tagged full-length recombinant human Cyclin-dependent kinase inhibitor 1 (p21).
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Purified
Human, Mouse
CDKN1A, CAP20, CDKN1, CIP1, MDA6, PIC1, SDI1, WAF1
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit