Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ p21 WAF1 Ab-3, Mouse Monoclonal Antibody

Achieve accurate, reproducible results with the Thermo Scientific™ p21WAF1 Ab-3, Mouse Monoclonal Antibody.

$145.20 - $667.50

Specifications

Antigen p21WAF1 Ab-3
Clone DCS-60.2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 7
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS230P
View Documents
Thermo Scientific™
MS230P
Purified Ab with BSA and Azide 1mL
Each for $632.00
Add to cart
 
MS230P0
View Documents
Thermo Scientific™
MS230P0
Purified Ab with BSA and Azide 100μL
Each for $145.20
Add to cart
 
MS230P1
View Documents
Thermo Scientific™
MS230P1
Purified Ab with BSA and Azide 500μL
Each for $413.00
Add to cart
 
MS230P1ABX
View Documents
Thermo Scientific™
MS230P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $470.00
Add to cart
 
MS230PABX
View Documents
Thermo Scientific™
MS230PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $667.50
Add to cart
 
MS230R7
View Documents
Thermo Scientific™
MS230R7
Ready-to-use for IHC (UVLP) 7mL
Each for $284.00
Add to cart
 
MS230R1
View Documents
Thermo Scientific™
MS230R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Achieve accurate, reproducible results with the Thermo Scientific™ p21WAF1 Ab-3, Mouse Monoclonal Antibody.

p21WAF1/Cip1/Sdi1/Pic1 is a tumor suppressor protein. Expression of p21WAF1 is induced by wild type, but not mutant, p53 suppressor protein. The p21WAF1 protein binds to cyclin/CDK complexes and inhibits their kinase activity thereby stopping cell cycle progression. It also binds to PCNA (proliferating cell nuclear antigen) and blocks DNA replication but not the DNA repair process.

Host Species: Mouse

Clone: DCS-60.2

Isotype: IgG2a

Species Reactivity: Human. Does not react with mouse and rat. Others not known

Epitope: Not determined

Immunogen: Full-length human recombinant p21 protein

Molecular Weight: 21kDa

Positive Control: Ls174T or Raji cells, Colon CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not suitable)
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry (Not suitable)

Specifications

Specifications

p21WAF1 Ab-3
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
IVD
Primary
IgG2a
DCS-60.2
Monoclonal
Murine
Cancer and Tumor Biology
Full length human recombinant p21 protein
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit