Learn More
MBL International Corporation P21/WAF1 100 U DFS Item
Manufacturer:  MBL International Corporation K00813
p21/WAF1 Monoclonal Antibody Size 100ug; Clone Number DCS-60 Host Species Mouse Conjugate Application WB, IPP, IHC
Catalog No. 50-481-11