Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

p23/PTGES3 Mouse anti-Human, Alexa Fluor 405, Clone: JJ6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11096879AF405

Catalog No. NBT96879405



Description

Description

p23/PTGES3 Monoclonal antibody specifically detects p23/PTGES3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

p23/PTGES3
JJ6
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
NP_006592.3
cPGESp23, Cytosolic prostaglandin E2 synthase, Hsp90 co-chaperone, P23cytosolic prostaglandin E synthase, Progesterone receptor complex p23, prostaglandin E synthase 3, prostaglandin E synthase 3 (cytosolic), TEBPEC 5.3.99.3, Telomerase-binding protein p23, unactive progesterone receptor, 23 kD
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Detects approx 23kDa.
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
Purified
PTGES3
Recombinant human full length p23 protein
Protein G purified
Membrane Trafficking and Chaperones
Primary
10728
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit