Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ p27 Kip1 Ab-1, Mouse Monoclonal Antibody

Achieve accurate, reproducible results with the Thermo Scientific™ p27Kip1 Ab-1, Mouse Monoclonal Antibody. 

Specifications

Antigen p27superscriptKip1superscript Ab-1
Clone DCS-72.F6
Applications Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 2
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity & Availability  
MS256P1ABX
View Documents
Thermo Scientific™
MS256P1ABX
1mg/mL Unconjugated 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS256P1
View Documents
Thermo Scientific™
MS256P1
200μg/mL Unconjugated 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Achieve accurate, reproducible results with the Thermo Scientific™ p27Kip1 Ab-1, Mouse Monoclonal Antibody.

p27Kip1, a candidate tumor suppressor gene, functions as a negative regulator of G1 progression and has been proposed to function as a possible mediator of TGF-Β induced G1 arrest.

Host Species: Mouse

Clone: DCS-72.F6

Isotype: IgG1/κ

Species Reactivity: Human, Mouse, Rat, and Dog. Does not react with cow. Others not known.

Epitope: aa 83-197

Immunogen: Mouse recombinant p27 protein

Molecular Weight: 27kDa

Positive Control: MAD109, Ls174T cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

p27superscriptKip1superscript Ab-1
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot
Murine
Cancer and Tumor Biology
Mouse recombinant p27 protein
DCS-72.F6
Monoclonal
RUO
Primary
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit