Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2RY8 Mouse anti-Human, Clone:1G5A11, Abnova™
Mouse monoclonal antibody raised against recombinant human P2RY8.
Supplier: Abnova Corporation MAB17577
Description
The protein encoded by this gene belongs to the family of G-protein coupled receptors, that are preferentially activated by adenosine and uridine nucleotides. This gene is moderately expressed in undifferentiated HL60 cells, and is located on both chromosomes X and Y. [provided by RefSeq]Specifications
P2RY8 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Immunocytochemistry (1:100-1:500) Flow Cytometry (1:200-1:400) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. |
|
P2RY8 | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG2a |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1G5A11 | |
purinergic receptor P2Y, G-protein coupled, 8 | |
In PBS (0.05% sodium azide) | |
MGC50878, P2Y8 | |
Recombinant protein corresponding to amino acid of human P2RY8 from E. coli. | |
286530 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction