Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2X1 Rabbit anti-Rat, Polyclonal, Neuromics™

Rabbit Polyclonal Antibody

Manufacturer:  Neuromics, IncSupplier Diversity Partner RA1010750UL

Encompass

 View more versions of this product

Catalog No. 50-210-0027


Add to cart

Description

Description

P2X1 Polyclonal antibody specifically detects P2X1 in Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Specifications

Specifications

P2X1
Polyclonal
Unconjugated
P2RX1
Liquid
Rabbit
IgG
RUO
Antibody
Polyclonal
Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Conc. Not Determined
whole serum with 0.05% sodium azide
P47824
AI323649; ATP receptor; BB122383; BOS_18726; P2rx1; P2x; P2X purinoceptor 1; P2X receptor, subunit 1; P2X1; P2X1 receptor; P2XMR; Pdcd3; programmed cell death 3; Purinergic receptor; purinergic receptor P2X 1; purinergic receptor P2X, ligand gated ion channel, 1; purinergic receptor P2X, ligand-gated ion channel, 1; purinergic receptor P2X1; purinergic receptor P2X1-1; purinergic receptor P2X1-2; RP-2; RP-2 protein
GLQENMRTS
50 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
25505
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit