Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2X2 Guinea Pig anti-Human, Rat, Polyclonal, Neuromics™ Non-distribution product as customer accommodation.

Guinea Pig Polyclonal Antibody

$200.88 - $448.88

Specifications

Antigen P2X2
Concentration Conc. Not Determined
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-0013 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
GP14106150UL
150 μL
Each for $448.88
Add to cart
 
50-210-0014 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
GP1410650UL
50 μL
Each for $200.88
Add to cart
 
Description

Description

P2X2 Polyclonal antibody specifically detects P2X2 in Human, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Specifications

Specifications

P2X2
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
Guinea Pig
Human, Rat
P49653, Q9UBL9
114115, 22953
IgG
Primary
Antibody
Conc. Not Determined
Polyclonal
Liquid
RUO
whole serum with 0.05% sodium azide
ATP receptor; BOS_16431; DFNA41; MGC129601; P2rx2; P2X purinoceptor 2; P2X Receptor, subunit 2; P2x2; P2X2 purinoceptor; P2X2 receptor splice variant P2X2-1; P2X2 receptor splice variant P2X2-2; P2X2 receptor splice variant P2X2-3; P2X2a; Purinergic receptor; purinergic receptor P2X 2; purinergic receptor P2X, ligand gated ion channel, 2; purinergic receptor P2X, ligand-gated ion channel, 2; purinergic receptor P2X2
DSTSTDPKGLAQL
Polyclonal
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
P2RX2
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit