Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ P2X3 Polyclonal Antibody

Guinea Pig Polyclonal Antibody

Supplier:  Invitrogen™ PA127969

Catalog No. PA127969


Only null left
Add to Cart

Description

Description

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel and may transduce ATP-evoked nociceptor activation. Mouse studies suggest that this receptor is important for peripheral pain responses, and also participates in pathways controlling urinary bladder volume reflexes. It is possible that the development of selective antagonists for this receptor may have a therapeutic potential in pain relief and in the treatment of disorders of urine storage.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

P2X3
Polyclonal
Unconjugated
P2rx3
4930513E20Rik; ATP receptor; MGC129956; P2RX3; P2X purinoceptor 3; p2X receptor 3; P2X receptor, subunit 3; P2X3; p2X3 purinoceptor; p2xr3; Purinergic receptor; purinergic receptor P2X 3; purinergic receptor P2X, ligand gated ion channel, 3; purinergic receptor P2X, ligand-gated ion channel, 3; purinergic receptor P2X3; purinoceptor P2X3
Guinea Pig
50 μL
Primary
Rat
Antibody
IgG
Immunohistochemistry (Frozen)
Conc. Not Determined
whole serum with 0.05% sodium azide
P49654
P2rx3
synthetic peptide VEKQSTDSGAYSIGH, corresponding to amino acids 383-397 of Rat P2X3.
RUO
81739
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.