Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2X3 Rabbit anti-Human, Non-human Primate, Rat, Polyclonal, Neuromics™ Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

$200.88 - $463.76

Specifications

Antigen P2X3
Concentration Conc. Not Determined
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-0030 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
RA10109150UL
150 μL
Each for $463.76
Add to cart
 
50-210-0031 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
RA1010950UL
50 μL
Each for $200.88
Add to cart
 
Description

Description

P2X3 Polyclonal antibody specifically detects P2X3 in Human, Non-human Primate, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Specifications

Specifications

P2X3
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Unconjugated
Rabbit
Human, Non-human Primate, Rat
P49654, P56373
5024, 81739
IgG
Primary
Antibody
Conc. Not Determined
Polyclonal
Liquid
RUO
whole serum with 0.05% sodium azide
4930513E20Rik; ATP receptor; MGC129956; P2RX3; P2X purinoceptor 3; p2X receptor 3; P2X receptor, subunit 3; P2X3; p2X3 purinoceptor; p2xr3; Purinergic receptor; purinergic receptor P2X 3; purinergic receptor P2X, ligand gated ion channel, 3; purinergic receptor P2X, ligand-gated ion channel, 3; purinergic receptor P2X3; purinoceptor P2X3
VEKQSTDSGAYSIGH
Polyclonal
-20° C, Avoid Freeze/Thaw Cycles
P2rx3
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit