Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2X6/P2RX6 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | P2X6/P2RX6 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:300-1:2000, Immunohistochemistry-Frozen 1:300-1:2000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP146608
|
Novus Biologicals
NBP146608 |
0.1 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB018635
|
Novus Biologicals
NBP1466080.025ML |
0.025 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
P2X6/P2RX6 Polyclonal antibody specifically detects P2X6/P2RX6 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
P2X6/P2RX6 | |
Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:300-1:2000, Immunohistochemistry-Frozen 1:300-1:2000 | |
Polyclonal | |
Antisera | |
RUO | |
Whole antisera with No Preservative | |
ATP receptor, MGC129625, P2RXL1skeletal muscle-expressed P2X, P2X purinoceptor 6, P2X6purinoceptor P2X6, P2XMpurinoreceptor P2X6, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 6, Purinergic receptor P2X-like 1, purinergic receptor P2X-like 1, orphan receptor | |
P2RX6 | |
IgG | |
Unpurified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
LYOPH | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Rabbit | |
Mouse, Rat | |
O54803 | |
9127 | |
A synthetic peptide from the cytoplasmic domain of mouse P2X6 (P2RX6, P2X purinoceptor 6, ATP receptor, P2RXL1) conjugated to an immunogenic carrier protein was used as the immunogen. | |
Primary | |
Reconstitute with 0.1 ml sterilized water to desired concentration. | |
Mouse, rat. Predicted cross-reactivity based on sequence identity: Human (86%). |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title