Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ P2X6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555975
Description
Immunogen sequence: HFYWRTKYEE AKAPKATANS VWRELALASQ ARLAECLRRS SAPAPTATAA GSQTQTPGWP CPSSDTHLPT HS Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 32%.
P2rx6 encodes receptors that are members of the ligand-gated ion channel family that open in response to extracellular ATP. Each P2rx6 receptor is made up of a trimer of subunits (P2X1-7) all of which share the common structure of two transmembrane domains. The protein encoded by the P2rx6 gene belongs to the family of P2X receptors, which are ATP-gated ion channels and mediate rapid and selective permeability to cations. The P2rx6 gene is predominantly expressed in skeletal muscle, and regulated by p53. The encoded protein is associated with VE-cadherin at the adherens junctions of human umbilical vein endothelial cells. Alternative splicing results in multiple transcript variants of P2rx6.
Specifications
P2X6 | |
Polyclonal | |
Unconjugated | |
P2RX6 | |
ATP receptor; P2RX6; P2RXL1; P2X purinoceptor 6; P2x6; P2X6 receptor; P2XM; Purinergic receptor; purinergic receptor P2X 6; purinergic receptor P2X, ligand gated ion channel, 6; purinergic receptor P2X, ligand-gated ion channel, 6; purinergic receptor P2X6; Purinergic receptor P2X-like 1; purinergic receptor P2X-like 1 orphan receptor; purinergic receptor P2X-like 1, orphan receptor; purinoceptor P2X6; purinoreceptor P2X6; Rnap2x6; skeletal muscle-expressed P2X | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9127 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15547 | |
P2RX6 | |
Recombinant protein corresponding to Human P2RX6. Recombinant protein control fragment (Product #RP-95342). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction