Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2X7/P2RX7 Antibody (Hano43) - BSA Free, Novus Biologicals™
SDP

Rat Monoclonal Antibody has been used in 2 publications

$206.50 - $517.59

Specifications

Antigen P2X7/P2RX7
Clone Hano43
Concentration 1 mg/mL
Dilution Flow Cytometry 1:25-1:200, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Applications Immunocytochemistry, Flow Cytometry, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018511
SDP
View Documents
Novus Biologicals
NBP1408940.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP14089401
SDP
View Documents
Novus Biologicals
NBP1408940.1MG
0.1 mg
Each for $517.59
Only null left
Add to Cart
 
Description

Description

P2X7/P2RX7 Monoclonal specifically detects P2X7/P2RX7 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

P2X7/P2RX7
1 mg/mL
Immunocytochemistry, Flow Cytometry, Immunofluorescence
Unconjugated
Rat
Mouse
Q9Z1M0
5027
A P2X7-expression construct and a final boost with P2X7-transfected HEK cells.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Hano43
Flow Cytometry 1:25-1:200, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Monoclonal
Purified
RUO
PBS (pH 7.4) with 0.09% Sodium Azide
ATP receptor, MGC20089, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A
P2RX7
IgG2b
Protein G purified
This antibody recognises the P2X purinoceptor 7, also known as P2X7. The P2X7 ATP receptor has a distinctive long C-terminal tail with multiple potential protein and lipid interaction motifs and is highly polymorphic. It is a cation selective ion channel that opens up on binding of extracellular ATP. Sustained activation by extracellular ATP results in the formation of a reversible pore in the plasma membrane that is permeable to hydrophilic solutes of up to 900 Da. Once a pore is opened massive upset of cytoplasmic ion homeostasis occurs and the pore stays open as long as it is bound by ATP. Should ATP stimulation continue the cell will become irreversibly damaged and die. P2X7 plays a key role in the maturation and release of IL-1 and other IL-1 family members during inflammation. As such, P2X7 blockers might be useful as anti-inflammatory agents.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.