Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P2Y1/P2RY1 Antibody (4G5D6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$548.00
Specifications
Antigen | P2Y1/P2RY1 |
---|---|
Clone | 4G5D6 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61664
![]() |
Novus Biologicals
NBP261664 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
P2Y1/P2RY1 Monoclonal specifically detects P2Y1/P2RY1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
P2Y1/P2RY1 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human, Rat | |
5028 | |
Purified recombinant fragment of human P2RY1 (AA: extra mix) expressed in E. Coli. | |
Primary | |
42 kDa |
4G5D6 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
Monoclonal | |
Purified | |
Apoptosis, Cytokine Research, GPCR, Plasma Membrane Markers | |
ATP receptor, P2 purinoceptor subtype Y1, P2Y purinoceptor 1, P2Y1platelet ADP receptor, Purinergic receptor, purinergic receptor P2Y, G-protein coupled, 1 | |
P2RY1 | |
IgG2b | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title