Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2Y1/P2RY1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $487.50

Specifications

Antigen P2Y1/P2RY1
Concentration LYOPH
Applications Immunohistochemistry (Frozen), Immunohistochemistry, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018501
SDP
View Documents
Novus Biologicals
NBP1307410.025ML
0.025 mL
Each for $181.90
Only null left
Add to Cart
 
NBP130741
SDP
View Documents
Novus Biologicals
NBP130741
0.1 mL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

P2Y1/P2RY1 Polyclonal specifically detects P2Y1/P2RY1 in Human, Mouse, Rat, Guinea Pig samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

P2Y1/P2RY1
Immunohistochemistry (Frozen), Immunohistochemistry, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Apoptosis, Cytokine Research, GPCR, Plasma Membrane Markers
P49650
5028
IgG
Unpurified
Appears to be specific for P2RY1.
LYOPH
Polyclonal
Antisera
RUO
Mouse, Human, Guinea Pig, Rat
ATP receptor, P2 purinoceptor subtype Y1, P2Y purinoceptor 1, P2Y1platelet ADP receptor, Purinergic receptor, purinergic receptor P2Y, G-protein coupled, 1
P2RY1
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.