Learn More
P2Y10/P2RY10 Mouse anti-Human, Alexa Fluor™ 405, Clone: 639329, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB10158V100UG
Catalog No. FB10158V100
Add to cart
This item is not returnable. View return policy
Description
Description
P2Y10/P2RY10 Monoclonal antibody specifically detects P2Y10/P2RY10 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
P2Y10/P2RY10
639329
Flow Cytometry 0.25-1 μg/106 cells
O00398
G-protein coupled purinergic receptor P2Y10, P2RY10, P2Y purinoceptor 10, P2Y10P2Y-like receptor, purinergic receptor P2Y, G-protein coupled, 10, putative P2Y purinoceptor 10
NS0 mouse myeloma cell line transfected with human P2RY10, Accession # O00398
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human P2RY10 in Flow Cytometry and Immunohistochemistry.
Flow Cytometry
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Mouse
IgG2b
100 μg
Primary
27334
Human