Learn More
P2Y11/P2RY11 Mouse anti-Human, Alexa Fluor 350, Clone: 505214, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB9305U100UG
Catalog No. FAB9305U100
Add to cart
This item is not returnable. View return policy
Description
Description
P2Y11/P2RY11 Monoclonal antibody specifically detects P2Y11/P2RY11 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
P2Y11/P2RY11
Monoclonal
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
P2RY11, P2Y11P2Y purinoceptor 11, purinergic receptor P2Y, G-protein coupled, 11, purinergic receptor P2Y11
NS0 mouse myeloma cell line transfected with human P2Y11/P2RY11 Accession # Q96G91
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Stains human P2Y11/P2RY11 transfectants but not irrelevant transfectants in flow cytometry.
Flow Cytometry
505214
Flow Cytometry 0.25-1 ug/10^6 cells
Q96G91
Mouse
IgG1
RUO
Primary
5032
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)