Learn More
P2Y13/P2RY13/GPR86 Mouse anti-Human, DyLight 488, Clone: 2H1G9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237382G
Catalog No. NBP237382G
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
P2Y13/P2RY13/GPR86
2H1G9
Western Blot, Flow Cytometry, ELISA
Purified
P2RY13
Purified recombinant fragment of human P2RY13 (AA: 1-49) expressed in E. Coli.
Protein A or G purified
GPCR
Primary
53829
Western Blot, Flow Cytometry, ELISA
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
G protein-coupled receptor 86, GPR86GPCR1, GPR94, G-protein coupled receptor 86, G-protein coupled receptor 94, P2Y purinoceptor 13, P2Y13FKSG77, purinergic receptor P2Y, G-protein coupled, 13, SP174
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human