Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2Y13/P2RY13/GPR86 Mouse anti-Human, FITC, Clone: 2H1G9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237382F

Catalog No. NBP237382F

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

P2Y13/P2RY13/GPR86
2H1G9
Flow Cytometry, ELISA
Purified
P2RY13
Purified recombinant fragment of human P2RY13 (AA: 1-49) expressed in E. Coli.
Protein A or G purified
GPCR
Primary
53829
ELISA, Flow Cytometry
FITC
PBS with 0.05% Sodium Azide
G protein-coupled receptor 86, GPR86GPCR1, GPR94, G-protein coupled receptor 86, G-protein coupled receptor 94, P2Y purinoceptor 13, P2Y13FKSG77, purinergic receptor P2Y, G-protein coupled, 13, SP174
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit