Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p33MONOX Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP156955
Description
p33MONOX Polyclonal specifically detects p33MONOX in Human samples. It is validated for Western Blot.Specifications
p33MONOX | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
Brain-derived rescue factor p60MONOX, FLJ21022, hypothetical protein LOC57179, KIAA1191, p60MONOX | |
Rabbit | |
Affinity purified | |
RUO | |
57179 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q96A73 | |
KIAA1191 | |
Synthetic peptides corresponding to KIAA1191(KIAA1191) The peptide sequence was selected from the middle region of KIAA1191. Peptide sequence TKYLRVAEALHKLKLQSGEVTKEERQPASAQSTPSTTPHSSPKQRPRGWF. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Guinea pig: 78%; Rabbit: 78%. | |
Human, Rat, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction