Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p33MONOX Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | p33MONOX |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP156834
|
Novus Biologicals
NBP156834 |
100 μL |
Each of 1 for $436.00
|
|
Description
p33MONOX Polyclonal specifically detects p33MONOX in Human samples. It is validated for Western Blot.Specifications
p33MONOX | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
Brain-derived rescue factor p60MONOX, FLJ21022, hypothetical protein LOC57179, KIAA1191, p60MONOX | |
KIAA1191 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
Q96A73 | |
57179 | |
Synthetic peptides corresponding to KIAA1191(KIAA1191) The peptide sequence was selected from the middle region of KIAA1191. Peptide sequence TPHSSPKQRPRGWFTSGSSTALPGPNPSTMDSGSGDKDRNLSDKWSLFGP. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title