Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ p35 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564865
Description
Immunogen sequence: FITPANVVFLY MLCRDVISSE VGSDHELQAV LLTCLYLSYS YMGNEISYPL KPFLVESCKE AFWDRCLSVI NLMSSKMLQI NADPHYFTQV FSDLKNESGQ EDK Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
The protein encoded by this gene (p35) is a neuron-specific activator of cyclin-dependent kinase 5 (CDK5); the activation of CDK5 is required for proper development of the central nervous system. The p35 form of this protein is proteolytically cleaved by calpain, generating a p25 form. The cleavage of p35 into p25 results in relocalization of the protein from the cell periphery to nuclear and perinuclear regions. P25 deregulates CDK5 activity by prolonging its activation and changing its cellular location. The p25 form accumulates in the brain neurons of patients with Alzheimer's disease. This accumulation correlates with an increase in CDK5 kinase activity, and may lead to aberrantly phosphorylated forms of the microtubule-associated protein tau, which contributes to Alzheimer's disease.
Specifications
p35 | |
Polyclonal | |
Unconjugated | |
Cdk5r1 | |
23 kDa subunit of tau-protein kinase II; cdk5 activator; CDK5 activator 1; CDK5P35; Cdk5r; Cdk5r1; cyclin dependent kinase 5 regulatory subunit 1; cyclin-dependent kinase 5 activator 1; Cyclin-dependent kinase 5 activator 1, p25; Cyclin-dependent kinase 5 activator 1, p35; Cyclin-dependent kinase 5 regulatory subunit 1; cyclin-dependent kinase 5, regulatory subunit (p35) 1; cyclin-dependent kinase 5, regulatory subunit 1 (p35); D11Bwg0379e; NCK5A; neuronal CDK5 activator; p23; p25; P25/P35; p35; p35nck5a; regulatory partner for CDK5 kinase; tau protein kinase II 23 kDa subunit; tau protein kinase II 23kDa subunit; TPKII regulatory subunit | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8851 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15078 | |
Cdk5r1 | |
Recombinant Human p35. Recombinant protein control fragment (Product #RP-105673). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction