Learn More
SAPK4 Monoclonal Antibody (OTI12B12), Invitrogen™
Mouse Monoclonal Antibody
Supplier:  Thermo Scientific MA526206
Catalog No. PIMA526206
Add to Cart
Description
Description
SAPK4 Monoclonal antibody specifically detects SAPK4 in Canine, Human, Mouse, Non-human Primate samples. It is validated for Immunocytochemistry, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SAPK4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O15264, Q9Z1B7
MAPK13
Full length human recombinant protein of MAPK13 produced in HEK293T cell
100 μL
Primary
Canine, Human, Mouse, Non-human Primate
Antibody
IgG2b
Immunocytochemistry, Western Blot
OTI12B12
Unconjugated
MAPK13
im:7136778; MAP kinase 13; MAP kinase p38 delta; MAPK 13; Mapk13; MAPK-13; mitogen activated protein kinase 13; mitogen-activated protein kinase 13; Mitogen-activated protein kinase p38 delta; p38 delta MAP kinase; p38delta; PRKM13; SAPK/Erk/kinase 4; SAPK4; Serk4; Stress-activated protein kinase 4
Mouse
Affinity Chromatography
RUO
26415, 5603, 612821
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions