Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p38 Mouse anti-Human, Mouse, Rat, Primate, FITC, Clone: 10B11C8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237312F

Catalog No. NBP237312F

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

p38
10B11C8
ELISA, Immunohistochemistry
Purified
MAPK14
Purified recombinant fragment of human MAPK14 (AA: 299-360) expressed in E. Coli.
Protein A or G purified
Cancer, Cell Cycle and Replication, Immunology, Innate Immunity, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction
Primary
1432
ELISA, Immunohistochemistry
FITC
PBS with 0.05% Sodium Azide
CSAID-binding protein, Csaids binding protein, CSBP, CSBP1MAX-interacting protein 2, CSBP2, CSPB1, cytokine suppressive inflammatory drug binding protein, Cytokine suppressive inflammatory drug-binding protein, EC 2.7.11, EC 2.7.11.24, EXIP, MAP kinase 14, MAP kinase MXI2, MAP kinase p38 alpha, MAPK 14, mitogen-activated protein kinase 14, Mitogen-activated protein kinase p38 alpha, Mxi2, p38, p38 MAP kinase, p38 mitogen activated protein kinase, p38ALPHA, p38alpha Exip, PRKM14, PRKM15, RK, SAPK2A, stress-activated protein kinase 2A
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse, Primate, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit