Learn More
P3H2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 157231AP
Catalog No. 50-173-0433
Add to cart
Description
Description
This gene encodes a member of the prolyl 3-hydroxylase subfamily of 2-oxo-glutarate-dependent dioxygenases. These enzymes play a critical role in collagen chain assembly, stability and cross-linking by catalyzing post-translational 3-hydroxylation of proline residues. Mutations in this gene are associated with nonsyndromic severe myopia with cataract and vitreoretinal degeneration, and downregulation of this gene may play a role in breast cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
Specifications
P3H2
0.14 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8CG71, Q8IVL5
leprecan like 1, Leprecan like protein 1, LEPREL1, MLAT4, P3H2, Prolyl 3 hydroxylase 2
Rabbit
IgG
150 μL
-20°C
Primary
210530, 55214
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
P3H2
Liquid
P3H2
P3H2 Fusion Protein Ag8444
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse