Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P3H3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 160231AP150UL
Description
The protein encoded by this gene belongs to the leprecan family of proteoglycans, which function as collagen prolyl hydroxylases that are required for proper collagen biosynthesis, folding and assembly. This protein, like other family members, is thought to reside in the endoplasmic reticulum. Epigenetic inactivation of this gene is associated with breast and other cancers, suggesting that it may function as a tumor suppressor.Specifications
P3H3 | |
Polyclonal | |
Unconjugated | |
P3H3 | |
GRCB, HSU47926, leprecan like 2, Leprecan like protein 2, LEPREL2, P3H3, Prolyl 3 hydroxylase 3, Protein B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10536, 14789, 297595 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CG70, Q8IVL6 | |
P3H3 | |
P3H3 Fusion Protein Ag8850 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction