Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p41 Flagellin Mouse anti-Bacteria, Clone: 6802, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30752410UG

 View more versions of this product

Catalog No. NB122521


Add to cart

Description

Description

p41 Flagellin Monoclonal antibody specifically detects p41 Flagellin in Bacteria samples. It is validated for ELISA, Immunofluorescence
Specifications

Specifications

p41 Flagellin
6802
Unconjugated
10 mM PBS with 0.05% BSA
41 kDa antigen, fla, Flagellar filament 41 kDa core protein, Flagellin, p41
A crude sonicate of Borrelia spirochetes
Protein A or G purified
RUO
Primary
46847386
ELISA, Immunofluorescence
0.2 mg/ml
ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Purified
Mouse
IgG2a Kappa
100 μg
Store at 4°C.
Monoclonal
Bacteria
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit