Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p41 Flagellin Mouse anti-Bacteria, Clone: 6802, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen p41 Flagellin
Clone 6802
Concentration 0.2 mg/ml
Dilution ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Applications ELISA, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122522
View Documents
Novus Biologicals
NBP30752420UG
20 μg
Each for $138.37
Add to cart
 
NB122521
View Documents
Novus Biologicals
NBP30752410UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

p41 Flagellin Monoclonal antibody specifically detects p41 Flagellin in Bacteria samples. It is validated for ELISA, Immunofluorescence
Specifications

Specifications

p41 Flagellin
0.2 mg/ml
ELISA, Immunofluorescence
Purified
RUO
10 mM PBS with 0.05% BSA
46847386
IgG2a Kappa
Primary
Store at 4°C.
6802
ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Unconjugated
Mouse
Bacteria
41 kDa antigen, fla, Flagellar filament 41 kDa core protein, Flagellin, p41
A crude sonicate of Borrelia spirochetes
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit