Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p44 MAPK (ERK1) Rabbit anti-Human,Mouse,Rat, KHL, Clone: 13F10, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen p44 MAPK (ERK1)
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB061822MI
View Documents
MilliporeSigma
ZRB0618225UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB061824MI
View Documents
MilliporeSigma
ZRB061824X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

p44 MAPK (ERK1) Monoclonal antibody specifically detects p44 MAPK (ERK1) in Human,Mouse,Rat samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.
Specifications

Specifications

p44 MAPK (ERK1)
KHL
Rabbit
Mitogen-activated protein kinase 3;EC: 2.7.11.24;MAP kinase 3;MAPK 3;ERT2;Extracellular signal-regulated kinase 1;ERK-1;Insulin-stimulated MAP2 kinase;MAP kinase isoform p44;MNK1;Microtubule-associated protein 2 kinase;p44-ERK1
KLH-conjugated linear peptide corresponding to 35 amino acids from the C-terminal region of rat p44 MAPK (ERK1).
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Purified
Human, Mouse, Rat
Mapk3, Erk1, Prkm3
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit