Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
P4HA3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA589422
Description
Immunogen sequence: DPERAAARGD TFSALTSVAR ALAPERRLLG LLRRYLRGEE ARLRDLTRFY DKVLSLHEDS TTPVANPLLA FTLIKRLQSD WRNVVHSLEA S; Positive Samples: 293T; Cellular Location: Endoplasmic reticulum lumen
Catalyzes the post-translational formation of 4- hydroxyproline in -Xaa-Pro-Gly- sequences in collagens and other proteins.Specifications
P4HA3 | |
Polyclonal | |
Unconjugated | |
P4HA3 | |
P4ha3 | |
Recombit fusion protein containing a sequence corresponding to amino acids 20-110 of human P4HA3 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
1.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6W3E9, Q6W3F0, Q7Z4N8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
283208, 320452, 361612 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title