Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ p53 Ab-4, Mouse Monoclonal Antibody

Achieve accurate, reproducible results with the Thermo Scientific™ p53 Ab-4, Mouse Monoclonal Antibody.

Specifications

Antigen p53 Ab-4
Clone PAb 122
Applications Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
MS182P0
View Documents
Thermo Scientific™
MS182P0
200μg/mL 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS182P1
View Documents
Thermo Scientific™
MS182P1
200μg/mL 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Achieve accurate, reproducible results with the Thermo Scientific™ p53 Ab-4, Mouse Monoclonal Antibody.

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Host Species: Mouse

Clone: PAb 122

Isotype: IgG2b/κ

Species Reactivity: Human, Monkey, Mouse, Rat, and Hamster. Others not known.

Epitope: aa 370-378

Immunogen: SV40-transformed mouse B4 cells

Molecular Weight: 53kDa

Positive Control: MDA-231 cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

p53 Ab-4
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
Murine
Cancer and Tumor Biology
SV40-transformed mouse B4 cells
PAb 122
Monoclonal
Concentrated
RUO
Primary
IgG2b kappa
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit