Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ p53 Ab-5, Mouse Monoclonal Antibody

Achieve accurate, reproducible results with the Thermo Scientific™ p53 Ab-5, Mouse Monoclonal Antibody.

$182.00 - $741.00

Specifications

Antigen p53 Ab-5
Clone DO-7
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 10
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS186B
View Documents
Thermo Scientific™
MS186B
Biotinylated Ab with BSA and Azide 1mL
Each for $741.00
Add to cart
 
MS186B0
View Documents
Thermo Scientific™
MS186B0
Biotinylated Ab with BSA and Azide 100μL
Each for $204.00
Add to cart
 
MS186B1
View Documents
Thermo Scientific™
MS186B1
Biotinylated Ab with BSA and Azide 500μL
Each for $641.00
Add to cart
 
MS186P
View Documents
Thermo Scientific™
MS186P
Purified Ab with BSA and Azide 1mL
Each for $595.50
Add to cart
 
MS186P0
View Documents
Thermo Scientific™
MS186P0
Purified Ab with BSA and Azide 100μL
Each for $182.00
Add to cart
 
MS186P1
View Documents
Thermo Scientific™
MS186P1
Purified Ab with BSA and Azide 500μL
Each for $424.50
Add to cart
 
MS186P1ABX
View Documents
Thermo Scientific™
MS186P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $572.50
Add to cart
 
MS186PABX
View Documents
Thermo Scientific™
MS186PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $614.00
Add to cart
 
MS186RQ
View Documents
Thermo Scientific™
MS186RQ
Ready-to-use for IHC (Quanto) 12mL
Each for $313.00
Add to cart
 
MS186R7
View Documents
Thermo Scientific™
MS186R7
Ready-to-use for IHC (UVLP) 7mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Achieve accurate, reproducible results with the Thermo Scientific™ p53 Ab-5, Mouse Monoclonal Antibody.

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Host Species: Mouse

Clone: DO-7

Isotype: IgG2b/κ

Species Reactivity: Human, Monkey, Cow. Others not known.

Epitope: aa 37-45

Immunogen: Recombinant human wild-type p53 protein expressed in E. coli

Molecular Weight: 53kDa

Positive Control: SKBR-3 or MDA-231 cells, Colon CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)
  • Flow Cytometry

Specifications

Specifications

p53 Ab-5
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Murine
Cancer and Tumor Biology
Recombinant human wild-type p53 protein expressed in E. coli
DO-7
Monoclonal
IVD
Primary
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit