Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ p53 Ab-6, Mouse Monoclonal Antibody

Achieve accurate, reproducible results with the Thermo Scientific™ p53 Ab-6, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS187P0

 View more versions of this product

Catalog No. MS187P0

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Achieve accurate, reproducible results with the Thermo Scientific™ p53 Ab-6, Mouse Monoclonal Antibody.

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Host Species: Mouse

Clone: DO-1

Isotype: IgG2a

Species Reactivity: Human, Monkey and Cow. Reacts weakly with Mouse and Rat. Others not known.

Epitope: aa 11-25

Immunogen: Recombinant human wild-type p53 protein expressed in E. coli

Molecular Weight: 53kDa

Positive Control: SKBR-3 cells or MDA-231 cells, Colon CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry
  • Gel Supershift Assay

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

p53 Ab-6
Monoclonal
200μg/mL
Purified Ab with BSA and Azide
Murine
IgG2a
RUO
Primary
Monoclonal
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
DO-1
Unconjugated
Concentrated
Recombinant human wild-type p53 protein expressed in E. coli
100μL
Cell Cycle
Primary
Bovine, Human, Non-human Primate
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit