Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ p53 Ab-8, Mouse Monoclonal Antibody

Deliver accurate, reproducible results with the Thermo Scientific™ p53 Ab-8, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS738P0

 View more versions of this product

Catalog No. MS738P0


Add to cart

Description

Description

Deliver accurate, reproducible results with the Thermo Scientific™ p53 Ab-8, Mouse Monoclonal Antibody.

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Host Species: Mouse

Clone: DO-7 + BP53-12

Isotype: IgG2b + IgG2a

Species Reactivity: Human. Others not known.

Epitope: aa 37-45 and aa 20-25

Immunogen: Recombinant human p53 protein

Molecular Weight: 53kDa

Positive Control: MDA-231, SKBR-3 cells or Colon CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)
Specifications

Specifications

p53 Ab-8
Monoclonal
200μg/mL
Purified Ab with BSA and Azide
Murine
IgG2a IgG2b
IVD
Primary
Monoclonal
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
DO-7 + BP53-12
Unconjugated
Concentrated
Recombinant human p53 protein
100μL
Cancer and Tumor Biology
Primary
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit