Learn More
Enzo Life SciencesSupplier Diversity Partner IMMUNOSET p53/MDM2 complex ELISA development set (96 wells)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI960070
First ELISA development product for the measurement of the p53/MDM2 complex for apoptosis and autophagy research. The IMMUNOSET p53/MDM2 Complex ELISA development set contains sufficient reagents for the development of 5 x 96 well EIA plates which provides analysis of ~200 samples in duplicate. Save time, money, and sample by developing your own EIA kits. Simply coat your plates and go! This kit provides superior accuracy with fully quantitative results compared to Western blot analysis and has low reactivity with related molecules. Contents: Capture antibody, p53 Standard, MDM2 Standard, Detection antibody, SA-HRP. Application: ELISA, Colorimetric detection. Use/Stability: Store all components at 4°C, except standard at -20°C. Assay Time: Plate Coating - Overnight + 1 hour; Assay - 3 hours. Crossreactivity: This assay detects p53/MDM2 and does not show significant reactivity with human p21 or Bax.
Catalog No. 50-658-674