Learn More
p53 Mouse anti-Mouse, Alexa Fluor™ 532, Clone: PAb242, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP253113AF532
Catalog No. NP23113A32
Add to cart
Description
Description
p53 Monoclonal antibody specifically detects p53 in Mouse samples. It is validated for Western Blot,ELISA,Immunohistochemistry,Immunocytochemistry,Immunofluorescence.
Specifications
Specifications
p53
PAb242
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Mouse
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
TP53
SV40-transformed cell line BALB/c SVA31 E7
0.1 mL
Primary
7157