Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ p53 (Mutant), Mouse, Unlabeled, Clone: PAb240,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ OP29100UG
Description
Recognizes the ∽53kDa mutant p53 protein under non-denaturing conditions by immunoprecipitation, immunofluorescence, and flow cytometry. Recognizes both mutant and wild-type p53 by immunoblotting and paraffin sections under denaturing conditions.Specifications
p53 (Mutant) | |
Monoclonal | |
Please refer to vial label for lot-specific concentration | |
In 50mM sodium phosphate buffer, 0.2% gelatin, pH 7.5, ≤0.1% Sodium Azide. | |
a recombinant protein consisting of amino acids 14-389 of p53 fused to β-galactosidase | |
RUO | |
Bovine | |
Liquid |
Gel Supershift, Immunofluorescence, Immunohistochemistry (Paraffin) | |
PAb240 | |
Unconjugated | |
Mouse | |
100 μg | |
Primary | |
2°-8°C | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Provide Content Correction