Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ p53 (Pantropic), Mouse, Unlabeled, Clone: DO-1,

Mouse Monoclonal Antibody

$136.00 - $408.00

Specifications

Antigen p53 (Pantropic)
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone DO-1
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
OP43100UG View Documents MilliporeSigma™
OP43100UG
100μg Unconjugated Each for $408.00
Add to cart
 
OP4320UG View Documents MilliporeSigma™
OP4320UG
20μg Unconjugated Each for $136.00
Add to cart
 
Description

Description

Recognizes the ~53kDa wild-type and mutant p53 protein in A431 cells and breast carcinoma tissue.
Specifications

Specifications

p53 (Pantropic)
Monoclonal
Mouse
In 0.05M sodium phosphate buffer, 0.2% gelatin, ≤0.1% Sodium Azide.
2°-8°C
Primary
DO-1
Liquid
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
RUO

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit